Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DIPA Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562965
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562965 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562965 Supplier Invitrogen™ Supplier No. PA562965
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ARQWQLFGTQ ASRAVREDLG GCWQKLAELE GRQEELLRE Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 38%.

DIPA functions as a transcriptional repressor. It may inhibit the activity of CTNNB1 in a TP53-dependent manner and thus regulate cell growth. It may function in adipocyte differentiation, negatively regulating mitotic clonal expansion.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DIPA
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CCDC85B
Gene Accession No. Q15834
Gene Alias AI842788; BB115872; CCDC85B; coiled-coil domain containing 85B; coiled-coil domain-containing protein 85B; Delta-interacting protein A; delta-interacting protein A homolog; Dipa; hepatitis delta antigen interacting protein A; hepatitis delta antigen-interacting protein A; Hepatitis delta antigen-interacting protein A homolog
Gene Symbols CCDC85B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DIPA. Recombinant protein control fragment (Product #RP-100766).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11007
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.