Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dipentaerythritol, 80+%, technical

Catalog No. p-121570
Change view
Click to view available options
Quantity:
25 g
500 g
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC117120250 25 g Plastic bottle
AC117125000 500 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC117120250 Supplier Thermo Scientific Chemicals Supplier No. 117120250
Only null left
Add to Cart

CAS: 126-58-9 | C10H22O7 | 254.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 126-58-9
Molecular Formula C10H22O7
Molecular Weight (g/mol) 254.28
MDL Number MFCD00004691
InChI Key TXBCBTDQIULDIA-UHFFFAOYSA-N
Synonym dipentaerythritol, dipentek, 2,2'-oxybis methylene bis 2-hydroxymethyl propane-1,3-diol, bis pentaerythritol, 1,3-propanediol, 2,2'-oxybis methylene bis 2-hydroxymethyl, 2,2,2',2'-tetrakis hydroxymethyl-3,3'-oxydipropan-1-ol, 1,3-propanediol, 2,2'-oxybis-methylene, 2-3-hydroxy-2,2-bis hydroxymethyl propoxy methyl-2-hydroxymethyl propane-1,3-diol
PubChem CID 31352
IUPAC Name 2-[[3-hydroxy-2,2-bis(hydroxymethyl)propoxy]methyl]-2-(hydroxymethyl)propane-1,3-diol
SMILES C(C(CO)(CO)COCC(CO)(CO)CO)O

Specifications

Melting Point 215°C to 225°C
Color White
Boiling Point 356°C
Flash Point >150°C
Infrared Spectrum Authentic
Assay Percent Range 85% min. (GC)
Packaging Plastic bottle
Linear Formula (HOCH2)3CCH2OCH2C(CH2OH)3
Quantity 25 g
Solubility Information Solubility in water: 2g/L (20°C). Other solubilities: 0.99g/100 mL water (50°C),2.1g/L methanol (25°C),0.6g/L ethanol (25°C)
Formula Weight 254.28
Percent Purity 85+%
Grade Technical
Physical Form Crystalline Powder
Chemical Name or Material Dipentaerythritol, 90%
Show More Show Less
Hazard Category HYGROSCOPIC
EINECSNumber 204-794-1
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.