Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dipentene, tech., Thermo Scientific Chemicals

Catalog No. p-7059959
Click to view available options
Quantity:
250 mL

Dipentene is a monoterpene olefin having potential applications in polymer and fuel chemistry. It is also widely used as flavoring and fragrance agent. It is a promising green solvent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 138-86-3
Molecular Formula C10H16
Molecular Weight (g/mol) 136.24
MDL Number MFCD00062992
InChI Key XMGQYMWWDOXHJM-UHFFFAOYNA-N
Synonym dipentene, limonene, cinene, cajeputene, dl-limonene, kautschin, p-mentha-1,8-diene, dipenten, polylimonene, eulimen
PubChem CID 22311
ChEBI CHEBI:15384
SMILES CC(=C)C1CCC(C)=CC1

Specifications

Density 0.85 g/mL
Boiling Point 170°C to 180°C
Flash Point 43°C (109°F)
Linear Formula CH3C6H8C(CH3)=CH2
Refractive Index 1.474
Quantity 250 mL
UN Number UN2052
Beilstein 3587825
Merck Index 14,5493
Solubility Information Soluble in ether (miscible) and ethanol (miscible). Insoluble in water.
IUPAC Name 1-methyl-4-prop-1-en-2-ylcyclohexene
Formula Weight 136.24
Grade Technical
Physical Form Liquid
Chemical Name or Material Dipentene
Show More Show Less
Hazard Category H226-H315-H317-H319
Hazard Statement GHS H Statement
H226-H315-H317
Flammable liquid and vapor.
Causes skin irritation.
May cause an allergic skin reaction.
Precautionary Statement P210-P233-P240-P241-P242-P243-P261-P264b-P272-P280-P303+P361+P353-P305+P351+P338-P333+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: DIPENTENE
EINECSNumber 205-341-0
RTECSNumber OS8100000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.