Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dipentyl phthalate, MilliporeSigma™ Supelco™

Catalog No. 111018537
Click to view available options
Quantity:
100 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Dipentyl phthalate belongs to the group of phthalates, widely used as plasticizers in plastics and for consumer applications. It is a major contaminant and can be released from a variety of products into the environment.

Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

Packaging Ampule
CAS 131-18-0
Density 1.025 g/mL (at 20°C)
Flash Point 118°C
Physical Form Neat
Linear Formula C18H26O4
MDL Number MFCD00041934
Type Analytical Standard
Quantity 100 mg
Formula Weight 306.40
Grade Analytical Standard
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.