Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diphenhydramine hydrochloride, 99%, Thermo Scientific Chemicals

Catalog No. AAA1013630
Click to view available options
Quantity:
50 g
250 g
1000 g

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A histamine H1 receptor antagonistDiphenhydramine hydrochloride acts as a sodium channel blocker. It is used as a histamine H1-receptor antagonist. Further, it is used as a sedative, hypnotic, antiemetic, antitussive and pruritus. It is employed in the treatment of allergic conditions. In addition to this, it is used as an active ingredient in common cold preparations.

Solubility
Soluble in water, dimethyl sulfoxide, dimethylformamide, methanol and ethanol.

Notes
Incompatible with strong oxidizing agents, strong acids and alkalis.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 147-24-0
Molecular Formula C17H22ClNO
Molecular Weight (g/mol) 291.82
MDL Number MFCD00012479
InChI Key PCHPORCSPXIHLZ-UHFFFAOYSA-N
Synonym diphenhydramine hydrochloride, dimedrol, diphenhydramine hcl, carphenamine, bendylate, carphenex, cathejell, denydryl, diphamine, eldadryl
PubChem CID 8980
ChEBI CHEBI:4637
IUPAC Name hydrogen [2-(diphenylmethoxy)ethyl]dimethylamine chloride
SMILES [H+].[Cl-].CN(C)CCOC(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 168°C to 169°C
Quantity 250 g
UN Number UN2811
Sensitivity Light sensitive
Solubility Information Soluble in water,dimethyl sulfoxide,dimethylformamide,methanol and ethanol.
Formula Weight 291.82
Percent Purity 99%
Chemical Name or Material Diphenhydramine hydrochloride
Hazard Category H302
Hazard Statement GHS H Statement
H351-H361-H302-H315-H319-H335
Suspected of causing cancer.
Suspected of damaging fertility or the unborn child.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 205-687-2
RTECSNumber KR7000000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.