Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diphenyl Diselenide 97.0+%, TCI America™
SDP

Catalog No. D218625G
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 1666-13-3
Molecular Formula C12H10Se2
Molecular Weight (g/mol) 312.15
MDL Number MFCD00003001
InChI Key YWWZCHLUQSHMCL-UHFFFAOYSA-N
Synonym diphenyl diselenide, diselenide, diphenyl, phenyl diselenide, 1,2-diphenyldiselane, diphenyldiselenide, bis phenylselenide, phenyldiselanyl benzene, diphenyldiselane, ccris 9227, acmc-209duq
PubChem CID 15460
IUPAC Name (phenyldiselanyl)benzene
SMILES [Se]([Se]C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 61°C
Color Yellow
Boiling Point 202°C
Quantity 25 g
UN Number 3283
Formula Weight 312.13
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Diphenyl Diselenide
RTECSNumber JM9152500
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.