Learn More
Diphenyl disulfide, 98%
Description
Diphenyl disulfide is one of the most popular organic disulfides used in organic synthesis. It is used as a reagent for the α-phenylsulfenylation of carbonyl compounds. It participates in hydrothiolation of alkynes via amine-mediated single electron transfer mechanism. It is the hydrolysis product of dyfonate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Diphenyl disulfide is one of the most popular organic disulfides used in organic synthesis. It is used as a reagent for the α-phenylsulfenylation of carbonyl compounds. It participates in hydrothiolation of alkynes via amine-mediated single electron transfer mechanism. It is the hydrolysis product of dyfonate.
Solubility
Insoluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
Specifications
| Melting Point | 58°C to 62°C |
| Boiling Point | 310°C |
| Odor | Unpleasant |
| Quantity | 50 g |
| Beilstein | 639794 |
| Solubility Information | Insoluble in water. |
| Formula Weight | 218.34 |
| Percent Purity | 98% |
| Chemical Name or Material | Diphenyl disulfide |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| DOTInformation | Hazard Class: 9; Packaging Group: III |
| EINECSNumber | 212-926-4 |
| RTECSNumber | SS6825000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.