Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bromonitromethane, 90%, Tech., Thermo Scientific Chemicals

Catalog No. AC307050010
Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 563-70-2
Molecular Formula CH2BrNO2
Molecular Weight (g/mol) 139.94
MDL Number MFCD00007401
InChI Key DNPRVXJGNANVCZ-UHFFFAOYSA-N
Synonym methane, bromonitro, bromo nitro methane, nitrobromomethane, unii-eg63p9eur0, eg63p9eur0, ccris 9456, bromo-nitromethane, 1-bromonitromethane, methane,bromonitro, monobromonitromethane
PubChem CID 68423
IUPAC Name bromo(nitro)methane
SMILES C([N+](=O)[O-])Br

Specifications

Color Yellow
Density 2.0700g/mL
Boiling Point 146°C to 148°C (750.0 mmHg)
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 88% min. (GC)
Packaging Glass bottle
Refractive Index 1.4850 to 1.495
Linear Formula BrCH2NO2
Quantity 1 g
Specific Gravity 2.07
Formula Weight 139.94
Percent Purity 90%
Grade Technical
Physical Form Liquid
Chemical Name or Material Bromonitromethane, tech., 90%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
May intensify fire; oxidizer.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Take any precaution to avoid mixing with combustibles.
In case of major fire and large quantities: Evacuate area.
Fight fire remotely due to the ris
EINECSNumber 209-258-0
RTECSNumber PA5360000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.