Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diphenyl Phenylphosphonate 98.0+%, TCI America™
SDP

Catalog No. D54541G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D54541G 1 g
D5454200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D54541G Supplier TCI America Supplier No. D54541G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3049-24-9
Molecular Formula C18H15O3P
Molecular Weight (g/mol) 310.29
MDL Number MFCD00216888
InChI Key CDOMXXVCZQOOMT-UHFFFAOYSA-N
Synonym Phenylphosphonic Acid Diphenyl Ester
PubChem CID 76448
IUPAC Name diphenyl phenylphosphonate
SMILES O=P(OC1=CC=CC=C1)(OC1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 75°C
Color White-Yellow
Boiling Point 165°C
Quantity 1 g
Formula Weight 310.29
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Diphenyl Phenylphosphonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.