Learn More
Diphenyl phosphorochloridate, 97%
Description
Diphenyl phosphorochloridate is used to prepare cyclohexyl-amidophosphoric acid diphenyl ester by reacting with cyclohexylamine. It is also used in the synthesis of alfa-substituted beta-lactams, anhydrides, esters and thioesters. It is also employed in the conversion of aldoximes to nitriles. Further, it is used in the preparation and reaction of enol phosphates, guanosine-adenosine-5',5'-triphosphate, guanosine-cytidine-5',5'-triphosphate, guanosine-uridine-5',5'-triphosphate, diguanosine-5',5'-diphosphate and diguanosine-5',5'-triphosphate. In addition to this, it serves as a phosphorylating agent for the preparation of 2-deoxy-D-galactose-3 and -6 phosphoric acids.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Diphenyl phosphorochloridate is used to prepare cyclohexyl-amidophosphoric acid diphenyl ester by reacting with cyclohexylamine. It is also used in the synthesis of alfa-substituted beta-lactams, anhydrides, esters and thioesters. It is also employed in the conversion of aldoximes to nitriles. Further, it is used in the preparation and reaction of enol phosphates, guanosine-adenosine-5′,5′-triphosphate, guanosine-cytidine-5′,5′-triphosphate, guanosine-uridine-5′,5′-triphosphate, diguanosine-5′,5′-diphosphate and diguanosine-5′,5′-triphosphate. In addition to this, it serves as a phosphorylating agent for the preparation of 2-deoxy-D-galactose-3 and -6 phosphoric acids.
Solubility
Immiscible with water.
Notes
Moisture sensitive. Incompatible with strong bases and strong oxidizing agents.
Specifications
Specifications
| Density | 1.296 |
| Boiling Point | 145°C to 147°C (1 mmHg) |
| Flash Point | 113°C (235°F) |
| Refractive Index | 1.55 |
| Quantity | 1000 g |
| UN Number | UN3265 |
| Beilstein | 654130 |
| Sensitivity | Moisture sensitive |
| Solubility Information | Immiscible with water. |
| Formula Weight | 268.64 |
| Show More |
Safety and Handling
| Hazard Category | H314 |
| Hazard Statement | GHS H Statement H314-H318 Causes severe skin burns and eye damage. Causes serious eye damage. |
| Precautionary Statement | P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c |
| DOTInformation | Hazard Class: 8; Packaging Group: II |
| EINECSNumber | 219-759-6 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.