Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diphenyl sulfoxide, 98+%

Catalog No. p-7026234
Click to view available options
Quantity:
25 g
100 g
500 g

CAS: 945-51-7 | C12H10OS | 202.271 g/mol

Diphenyl sulfoxide is a reagent used as an oxidant and hydroxyl activator in combination with a variety of electrophilic reagents. Efficient glycosidation of a phenyl thiosialoside donor with diphenyl sulfoxide and triflic anhydride in dichloromethane was reported. Direct glycosylations with 1-hydroxy glycosyl donors is made using trifluoromethanesulfonic anhydride and diphenyl sulfoxide. The preparation of triarylsulfonium halides was by the action of aryl Grignard reagents on diphenyl sulfoxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diphenyl sulfoxide is a reagent used as an oxidant and hydroxyl activator in combination with a variety of electrophilic reagents. Efficient glycosidation of a phenyl thiosialoside donor with diphenyl sulfoxide and triflic anhydride in dichloromethane was reported. Direct glycosylations with 1-hydroxy glycosyl donors is made using trifluoromethanesulfonic anhydride and diphenyl sulfoxide. The preparation of triarylsulfonium halides was by the action of aryl Grignard reagents on diphenyl sulfoxide.

Solubility
Soluble in water. Soluble in most organic solvents.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 945-51-7
Molecular Formula C12H10OS
Molecular Weight (g/mol) 202.271
MDL Number MFCD00002085
InChI Key JJHHIJFTHRNPIK-UHFFFAOYSA-N
Synonym diphenyl sulfoxide, phenyl sulfoxide, sulfinyldibenzene, phenylsulfinylbenzene, diphenyl sulphoxide, sulfoxide, diphenyl, diphenylsulfoxide, benzene, 1,1'-sulfinylbis, 1,1'-sulfinyldibenzene, phenylsulfinyl benzene
PubChem CID 13679
IUPAC Name benzenesulfinylbenzene
SMILES C1=CC=C(C=C1)S(=O)C2=CC=CC=C2

Specifications

Melting Point 69°C to 72°C
Boiling Point 206°C to 208°C (13 mmHg)
Quantity 25 g
Beilstein 1908444
Solubility Information Soluble in water. Soluble in most organic solvents.
Formula Weight 202.28
Percent Purity ≥98%
Chemical Name or Material Diphenyl sulfoxide
EINECSNumber 213-415-9
RTECSNumber DA9185000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.