Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diphenylacetic Acid, 99+%

Catalog No. p-122087
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Plastic bottle

CAS: 117-34-0 | C14H12O2 | 212.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 117-34-0
MDL Number MFCD00004251
InChI Key PYHXGXCGESYPCW-UHFFFAOYSA-N
Synonym diphenylacetic acid, diphenylethanoic acid, acetic acid, diphenyl, benzeneacetic acid, .alpha.-phenyl, 1,1-diphenylacetic acid, unii-658ncz0nko, diphenyl-acetic acid, alpha-toluic acid, alpha-phenyl, 658ncz0nko, benzeneacetic acid, alpha-phenyl
PubChem CID 8333
ChEBI CHEBI:41967
IUPAC Name 2,2-diphenylacetic acid
SMILES C1=CC=C(C=C1)C(C2=CC=CC=C2)C(=O)O

Specifications

Melting Point 146.0°C to 149.0°C
Color Cream-White to White
Loss on Drying 0.5% max.
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Molecular Formula C14H12O2
Linear Formula (C6H5)2CHCO2H
Quantity 5 g
Beilstein 09, 673
Merck Index 15, 3346
Solubility Information Solubility in water: soluble in hot water. Other solubilities: soluble in alcohol, ether and chloroform
Molecular Weight (g/mol) 212.25
Formula Weight 212.25
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material Diphenylacetic acid, 99+%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 204-185-0
RTECSNumber AH2515000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.