Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diphenylamine, 0.3% (w/v) in 50% (v/v) Sulfuric Acid, Ricca Chemical
SDP

Catalog No. p-3340501
Click to view available options
Quantity:
120 mL
16 oz.
1 L
Packaging:
Amber Glass Bottle
Safety-coated Glass Bottle

Chemical Identifiers

CAS 7664-93-9, 7732-18-5, 122-39-4
Molecular Formula C12H11N
Molecular Weight (g/mol) 169.23
InChI Key DMBHHRLKUKUOEG-UHFFFAOYSA-N
Synonym diphenylamine, n-phenylbenzenamine, anilinobenzene, benzenamine, n-phenyl, n,n-diphenylamine, phenylaniline, scaldip, n-phenylbenzeneamine, big dipper, no-scald
PubChem CID 11487
ChEBI CHEBI:4640
IUPAC Name N-phenylaniline
SMILES N(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Color Colorless
CAS Min % 0.15, 46.89, 51.0
CAS Max % 0.16, 48.8, 53.0
Packaging Amber Glass Bottle
Quantity 120 mL
Physical Form Liquid
Percent Purity 0.3% W/V
Grade Laboratory
Concentration or Composition (by Analyte or Components) 0.3% (w/v) in 50% (v/v) Sulfuric Acid
Chemical Name or Material Diphenylamine
ShelfLife 12 months
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.