Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diphenylamine, 1% w/v, Reagents

Catalog No. p-200021533
Click to view available options
Quantity:
120 mL
500 mL

Catalog No. 01-335-436

Supplier: Reagents Holdings Llc CD001900120A

Only null left
Add to Cart
Add to Cart

Diphenylamine, 1% w/v, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5, 6152-67-6
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym N-Phenylbenzeneamine, N-Phenylaniline, N,N-Diphenylamine
IUPAC Name water
SMILES O

Specifications

Boiling Range 100°C
Color Green
Boiling Point 100°C
Packaging Poly Bottle
Quantity 120 mL
Specific Gravity 1.005
Percent Purity 1%
Grade Reagent
Physical Form Liquid
Chemical Name or Material Diphenylamine, 1% w/v
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.