Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diphenylamine, 1% (w/v), Ricca Chemical
SDP

Catalog No. 258716 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
120 mL
500 mL
Packaging:
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
258716 500 mL Natural Poly Bottle
25874 120 mL Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 258716 Supplier Ricca Chemical Company Supplier No. 258716
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7732-18-5 , 6152-67-6
Molecular Formula C12H11NNaO3S
Molecular Weight (g/mol) Mixture
MDL Number MFCD00036468
InChI Key VMVSDVIYIMNZBS-UHFFFAOYSA-N
Synonym sodium 4-phenylamino benzenesulfonate, sodium diphenylamine-4-sulfonate, benzenesulfonic acid, 4-phenylamino-, monosodium salt, unii-3e2ux1382o, sodium 4-anilinobenzenesulfonate, diphenylamine-4-sulfonic acid sodium salt, sodium diphenylamine-p-sulfonate, n-phenylsulfanilic acid, sodium salt, 4-phenylamino benzenesulfonic acid sodium salt, diphenylaminesulfonic acid sodium salt
PubChem CID 517002
IUPAC Name 4-(phenylamino)benzene-1-sulfonic acid sodium
SMILES [Na].OS(=O)(=O)C1=CC=C(NC2=CC=CC=C2)C=C1

Specifications

CAS Min % 97.0, 0.97
CAS Max % 100.0, 1.01
Color Colorless
Boiling Point Approximately 100°C
Odor Odorless
Packaging Natural Poly Bottle
Quantity 500 mL
Percent Purity 1% w/v
Grade Laboratory
Concentration or Composition (by Analyte or Components) 1% (w/v) Aqueous
Physical Form Liquid
Chemical Name or Material Diphenylamine
Show More Show Less
ShelfLife 12 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.