Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diphenylamine, 98+%

Catalog No. AAA182650E
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA182650E 2500 g
AAA1826522 100 g
AAA1826536 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA182650E Supplier Thermo Scientific Chemicals Supplier No. A182650E
Add to Cart

CAS: 122-39-4 | C12H11N | 169.227 g/mol

Diphenylamine is employed as harvest scald inhibitor for apples indoor drench treatments due to its antioxidant property. Alkyl derivatives of diphenylamine are used as anti-ozonants in the rubber manufacturing. It is used as lubricants and acts as a stabilizer for plastics. In the analytical chemistry, it is used to test the nitrate ion in association with ammonium chloride in sulfuric acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diphenylamine is employed as harvest scald inhibitor for apples indoor drench treatments due to its antioxidant property. Alkyl derivatives of diphenylamine are used as anti-ozonants in the rubber manufacturing. It is used as lubricants and acts as a stabilizer for plastics. In the analytical chemistry, it is used to test the nitrate ion in association with ammonium chloride in sulfuric acid.

Solubility
Soluble in ethanol, acetone, benzene, carbon tetrachloride, pyridine and ethyl acetate. Slightly soluble in chloroform.

Notes
Incompatible with strong acids, strong oxidizing agents. Keep the container tightly closed in a cool and well-ventilated area.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122-39-4
Molecular Formula C12H11N
Molecular Weight (g/mol) 169.227
MDL Number MFCD00003014
InChI Key DMBHHRLKUKUOEG-UHFFFAOYSA-N
Synonym diphenylamine, n-phenylbenzenamine, anilinobenzene, benzenamine, n-phenyl, n,n-diphenylamine, phenylaniline, scaldip, n-phenylbenzeneamine, big dipper, no-scald
PubChem CID 11487
ChEBI CHEBI:4640
IUPAC Name N-phenylaniline
SMILES C1=CC=C(C=C1)NC2=CC=CC=C2

Specifications

Melting Point 52°C to 55°C
Density 1.16
Boiling Point 302°C
Flash Point 152°C (305°F)
Odor Weak
Linear Formula (C6H5)2NH
Quantity 2500 g
UN Number UN3077
Beilstein 508755
Sensitivity Air and light sensitive
Merck Index 14,3317
Solubility Information Soluble in ethanol,acetone,benzene,carbon tetrachloride,pyridine and ethyl acetate. Slightly soluble in chloroform.
Formula Weight 169.23
Percent Purity ≥98%
Chemical Name or Material Diphenylamine
Show More Show Less
Hazard Category H301+H311+H331-H373
Hazard Statement GHS H Statement
H301-H311-H331-H373
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement P260-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P311-P312-P314-P330-P361-P363-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber 204-539-4
RTECSNumber JJ7800000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.