Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diphenylbis(phenylethynyl)silane 98.0+%, TCI America™
SDP

Catalog No. D43121G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D43121G 1 g
D43125G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D43121G Supplier TCI America Supplier No. D43121G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 18784-61-7
Molecular Formula C28H20Si
Molecular Weight (g/mol) 384.553
MDL Number MFCD00093508
InChI Key BBVOORFIXBLFFX-UHFFFAOYSA-N
PubChem CID 5213428
IUPAC Name diphenyl-bis(2-phenylethynyl)silane
SMILES C1=CC=C(C=C1)C#C[Si](C#CC2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=CC=C4

Specifications

Melting Point 81°C
Color White
Quantity 1 g
Formula Weight 384.55
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Diphenylbis(phenylethynyl)silane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.