Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diphenylborinic anhydride, 95%

Catalog No. AAH5550903
Click to view available options
Quantity:
250 mg
1 g
5 g

CAS: 4426-21-5 | C24H20B2O | 346.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4426-21-5
Molecular Formula C24H20B2O
Molecular Weight (g/mol) 346.04
MDL Number MFCD00042663
InChI Key SNQFEECGHGUHBK-UHFFFAOYSA-N
Synonym diphenylborinic anhydride, oxybis diphenylborane, diphenylboranyloxy diphenyl borane, diphenylboronic anhydride, 1,1,3,3-tetraphenyldiboroxane, diphenylboranyl diphenylborinate, tetraphenyldiboroxane, borane, oxybis diphenyl, diphenyl borinic anhydride, d03twr
PubChem CID 596810
IUPAC Name diphenylboranyloxy(diphenyl)borane
SMILES O(B(C1=CC=CC=C1)C1=CC=CC=C1)B(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 135°C to 140°C
Color White to Yellow
Linear Formula (C6H5)2BOB(C6H5)2
Quantity 1 g
Beilstein 2950331
Sensitivity Moisture sensitive
Formula Weight 346.04
Percent Purity 95%
Physical Form Powder
Chemical Name or Material Diphenylborinic anhydride
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.