Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diphenylcarbamoyl Chloride 98.0+%, TCI America™
SDP

Catalog No. D088025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D088025G 25 g
D08805G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D088025G Supplier TCI America Supplier No. D088025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 83-01-2
Molecular Formula C13H10ClNO
Molecular Weight (g/mol) 231.679
MDL Number MFCD00000633
InChI Key XNBKKRFABABBPM-UHFFFAOYSA-N
Synonym diphenylcarbamyl chloride, diphenylcarbamoyl chloride, carbamic chloride, diphenyl, diphenylcarbamic chloride, n,n-diphenylcarbamyl chloride, diphenylchloroformamide, carbamoyl chloride, diphenyl, carabamic chloride, diphenyl, chloroformic acid diphenylamide, carbamic chloride, n,n-diphenyl
PubChem CID 65741
IUPAC Name N,N-diphenylcarbamoyl chloride
SMILES C1=CC=C(C=C1)N(C2=CC=CC=C2)C(=O)Cl

Specifications

Melting Point 83°C
Color Blue-White
UN Number 3261
Quantity 25 g
Formula Weight 231.68
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Diphenylcarbamoyl Chloride
RTECSNumber EY5065000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.