Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diphenylcarbazide, 0.5% (w/v) in Acetone, Ricca Chemical
SDP

Catalog No. 25904 Shop All Ricca Chemical Company Products
Click to view available options
Quantity:
120 mL
500 mL
Packaging:
Amber Glass Bottle

Chemical Identifiers

CAS 140-22-7, 67-64-1
Molecular Formula C13H14N4O
Molecular Weight (g/mol) 242.282
InChI Key KSPIHGBHKVISFI-UHFFFAOYSA-N
Synonym 1,5-diphenylcarbazide, diphenylcarbazide, 1,5-diphenylcarbohydrazide, carbonic dihydrazide, 2,2'-diphenyl, 1,5-diphenylcarbonohydrazide, 1,5-diphenylcabohydrazide, sym-diphenylcarbazide, n,n'-diphenylcarbazide, 2,2'-diphenylcarbazide, carbohydrazide, 1,5-diphenyl
PubChem CID 8789
ChEBI CHEBI:4641
IUPAC Name 1,3-dianilinourea
SMILES C1=CC=C(C=C1)NNC(=O)NNC2=CC=CC=C2

Specifications

Color Colorless to Yellow
CAS Min % 97.0, 0.62
CAS Max % 100.0, 0.64
Percent Purity 0.5%
Grade Laboratory
Packaging Amber Glass Bottle
Solvent or Matrix Acetone
Quantity 120 mL
Physical Form Liquid
Concentration or Composition Notes 0.5% (w/v)
Chemical Name or Material Diphenylcarbazide
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.