Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Diphenylcarbazone Indicator, 0.1% (w/v) Alcoholic Solution, Ricca Chemical
Shop All Ricca Chemical Company Products

Click to view available options
Quantity:
120 mL
500 mL
1 L
4 L
Packaging:
Amber Glass Bottle
Specifications
Specifications
| Chemical Name or Material | Diphenylcarbazone Indicator |
| CAS | 64-17-5, 67-56-1, 538-62-5, 140-22-7 |
| CAS Min % | 0.12, 98.0 |
| CAS Max % | 0.13, 100.0 |
| Concentration | 0.1% (w/v) |
| Molecular Formula | C13H12N4O |
| Synonym | 1,5-diphenylcarbazone, diazenecarboxylic acid, phenyl-, 2-phenylhydrazide, unii-5uyj4r0d5t, phenylazoformic acid 2-phenylhydrazide, sym-diphenylcarbazone, 5uyj4r0d5t, chembl79409, 3-hydroxy-1,5-diphenylformazan, phenyldiazenecarboxylic acid 2-phenylhydrazide, 3-phenylamino-1-phenylimino urea |
| InChI Key | ZFWAHZCOKGWUIT-UHFFFAOYSA-N |
| SMILES | O=C(NNC1=CC=CC=C1)N=NC1=CC=CC=C1 |
| IUPAC Name | 3-(phenylamino)-1-(phenylimino)urea |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction