Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diphenylcyclopropenone, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
886-38-4
C15H10O
206.24
HCIBTBXNLVOFER-UHFFFAOYSA-N
diphenylcyclopropenone,diphencyprone,2,3-diphenylcyclopropenone,2,3-diphenylcycloprop-2-enone,cyclopropenone, diphenyl,dpcp,2-cyclopropen-1-one, 2,3-diphenyl,1,2-diphenylcyclopropen-3-one,cyclopropenone, 2,3-diphenyl,unii-i7g14nw5ec
65057
CHEBI:53074
2,3-diphenylcycloprop-2-en-1-one
C1=CC=C(C=C1)C2=C(C2=O)C3=CC=CC=C3
886-38-4
C15H10O
206.24
HCIBTBXNLVOFER-UHFFFAOYSA-N
diphenylcyclopropenone,diphencyprone,2,3-diphenylcyclopropenone,2,3-diphenylcycloprop-2-enone,cyclopropenone, diphenyl,dpcp,2-cyclopropen-1-one, 2,3-diphenyl,1,2-diphenylcyclopropen-3-one,cyclopropenone, 2,3-diphenyl,unii-i7g14nw5ec
65057
CHEBI:53074
2,3-diphenylcycloprop-2-en-1-one
C1=CC=C(C=C1)C2=C(C2=O)C3=CC=CC=C3
Quantity 1g
Color Beige to Orange-Pink
Melting Point 117.0°C to 121.0°C
Infrared Spectrum Authentic
Packaging Glass bottle
Assay Percent Range 97.5% min. (HPLC)
Beilstein 07,IV,1736
Merck Index 15,3338
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in methanol, ethanol, acetone, chloroform,, dichloromethane, cyclohexane, benzene, hexane,, petroleum ether
Formula Weight 206.24
Physical Form Crystalline Powder
Percent Purity 98%
Chemical Name or Material Diphenylcyclopropenone
Show More Show Less

GHS H Statement
May cause an allergic skin reaction.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
IF IN EYES: Rinse cautious

GHS Signal Word: Warning

EINECSNumber : 212-948-4