Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diphenyliodonium chloride, 97%

Catalog No. AC117330500
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle

Catalog No. AC117330500

Supplier: Thermo Scientific Chemicals 117330500

Only null left
Add to Cart

CAS: 1483-72-3 | C12H10ClI | 316.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1483-72-3
Molecular Formula C12H10ClI
Molecular Weight (g/mol) 316.57
MDL Number MFCD00011909
InChI Key RSJLWBUYLGJOBD-UHFFFAOYSA-M
Synonym diphenyliodonium chloride, diphenyliodanium chloride, iodonium, diphenyl-, chloride, diphenyliodoniumchloride, c12h10i.cl, iodonium, diphenyl-, chloride 1:1, acmc-1bxx9
PubChem CID 73870
IUPAC Name diphenyliodanium;chloride
SMILES [Cl-].[I+](C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 227°C to 228°C
Color Cream-Yellow to White
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula (C6H5)2ICl
Quantity 50 g
Fieser 01,341; 02,178
Solubility Information Solubility in water: soluble. Other solubilities: soluble in methanol
Formula Weight 316.57
Percent Purity 97%
Physical Form Powder
Chemical Name or Material Diphenyliodonium chloride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
If skin irritation occurs: Get medical advice/attention.
If eye irritatio
EINECSNumber 216-049-8

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.