Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diphenylmethane 99%, ACROS Organics™

Click to view available options
Quantity:
500g
1kg
Packaging:
Glass bottle
101-81-5
C13H12
168.24
MFCD00004781
CZZYITDELCSZES-UHFFFAOYSA-N
diphenylmethane,ditan,ditane,benzene, 1,1'-methylenebis,diphenyl methane,benzene, benzyl,methane, diphenyl,1,1'-dimethylenebis benzene,benzene, phenylmethyl,methylenedibenzene
7580
CHEBI:38884
benzylbenzene
C1=CC=C(C=C1)CC2=CC=CC=C2
101-81-5
C13H12
168.24
MFCD00004781
CZZYITDELCSZES-UHFFFAOYSA-N
diphenylmethane,ditan,ditane,benzene, 1,1'-methylenebis,diphenyl methane,benzene, benzyl,methane, diphenyl,1,1'-dimethylenebis benzene,benzene, phenylmethyl,methylenedibenzene
7580
CHEBI:38884
benzylbenzene
C1=CC=C(C=C1)CC2=CC=CC=C2
Color Colorless to Yellow
Melting Point 24.5°C to 25.0°C
Specific Gravity 1
Infrared Spectrum Authentic
Quantity 500g
Boiling Point 265.0°C
Refractive Index 1.5760 to 1.5780
Density 1.0000g/mL
Packaging Glass bottle
Flash Point 127°C
Assay Percent Range 99%
Linear Formula (C6H5)2CH2
Beilstein 05, II, 498
Merck Index 15, 3362
Solubility Information Solubility in water: practically insoluble. Other solubilities: insoluble in liquid ammonia, freely soluble in alcohol, ether, chloroform,, hexane and benzene
Formula Weight 168.24
Physical Form Low Melting Solid
Percent Purity 99%
Chemical Name or Material Diphenylmethane, 99%
Show More Show Less

EINECSNumber : 202-978-6

RTECSNumber : DA4976500

TSCA : TSCA