Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diphenylphosphine

Catalog No. p-7080296
Click to view available options
Quantity:
10 g

CAS: 829-85-6 | C12H11P | 186.19 g/mol

Diphenylphosphine acts as an intermediate in the preparation of diphenylphosphide derivatives, phosphonium salts, phosphine ligands and Wittig-Horner reagents. The presence of hydrogen atom bonded to phosphorus undergoes Michael-like addition to activated alkenes. It is involved in the preparation of 1,2-bis(diphenylphosphino)ethane and (phenyl-(phenylmethyl)phosphoryl)benzene. Further, it is used in the synthesis of aminophosphines and chiral palladacycles with N-heterocyclic carbene ligands as catalysts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 829-85-6
Molecular Formula C12H11P
Molecular Weight (g/mol) 186.19
MDL Number MFCD00003040
InChI Key GPAYUJZHTULNBE-UHFFFAOYSA-N
Synonym diphenylphosphine, phosphine, diphenyl, ph2ph, unii-f9b5t7o7zy, diphenyl phosphine, hpph2, f9b5t7o7zy, diphenylphosphine 10wt%inhexane, diphenylphospine, diphenyl-phosphine
PubChem CID 70017
IUPAC Name diphenylphosphane
SMILES P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Density 1.07 g/mL
Boiling Point 280°C
Flash Point 110°C (230°F)
Quantity 10 g
UN Number UN2845
Sensitivity Air, light, and moisture sensitive
Solubility Information Miscible with ethanol,ether,benzene,concentrated hydrochloric acid. Immiscible with water.
Formula Weight 186.2
Physical Form Liquid
Chemical Name or Material Diphenylphosphine
Hazard Category H250-H302-H315-H319-H335
Hazard Statement GHS H Statement
H250-H252-H315-H319-H335
Precautionary Statement P210-P222-P261-P264b-P270-P271-P280-P301+P312-P302+P334-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.2; Packing Group: I; Proper Shipping Name: PYROPHORIC LIQUIDS, ORGANIC, N.O.S.
EINECSNumber 212-591-4
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.