Diphenylphosphine

Manufacturer: Alfa Aesar 5616909

Quantity 10g

 View more versions of this product

Catalog No. AA5616909

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 829-85-6
    Molecular Formula (C6H5)2PH
    Formula Weight 186.2
    MDL Number MFCD00003040
    UN Number UN2845
    Chemical Name or Material Diphenylphosphine

    Specifications

    Formula Weight 186.2
    Quantity 10g
    UN Number UN2845
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H250-H252-H315-H319-H335 Catches fire spontaneously if exposed to air. Self-heating in large quantities may catch fire. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P210-P222-P305+P351+P338-P405-P422a-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Do not allow contact with air.
    IF IN EYES: Rinse cautiously with water for several minutes. Re
    DOT Information Transport Hazard Class: 4.2; Packing Group: I; Proper Shipping Name: PYROPHORIC LIQUIDS, ORGANIC, N.O.S.
    Physical Form Liquid
    Density 1.07 g/mL
    Molecular Formula (C6H5)2PH
    MDL Number MFCD00003040
    CAS 829-85-6
    Chemical Name or Material Diphenylphosphine

    GHS H Statement
    H250-H252-H315-H319-H335 Catches fire spontaneously if exposed to air. Self-heating in large quantities may catch fire. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

    GHS P Statement
    P210-P222-P305+P351+P338-P405-P422a-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Do not allow contact with air.
    IF IN EYES: Rinse cautiously with water for several minutes. Re

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit