Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rhodium(III) acetylacetonate, 97%

Catalog No. AC325450050
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle

CAS: 14284-92-5 | C15H21O6Rh | 400.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14284-92-5
Molecular Formula C15H21O6Rh
Molecular Weight (g/mol) 400.23
MDL Number MFCD00083144 MFCD00083144
InChI Key DGOINFUDFBWCMX-LNTINUHCSA-K
Synonym rhodium iii acetylacetonate-rh
PubChem CID 131675882
IUPAC Name (E)-4-hydroxypent-3-en-2-one;rhodium
SMILES [Rh+3].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O

Specifications

Melting Point 263.0°C to 264.0°C
Color Orange-Yellow to Yellow
Infrared Spectrum Authentic
Assay Percent Range 24.7% min. (Rh)
Packaging Glass bottle
Linear Formula (CH3COCH2=C(O-)CH3)3Rh
Quantity 5 g
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ethyl ether, ethanol
Formula Weight 400.24
Percent Purity 97%
Physical Form Powder
Chemical Name or Material Rhodium(III) acetylacetonate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 238-192-5

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.