Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diphenylphosphinic acid, 99%

Catalog No. p-123006
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle

CAS: 1707-03-5 | C12H11O2P | 218.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1707-03-5
Molecular Formula C12H11O2P
Molecular Weight (g/mol) 218.19
MDL Number MFCD00002132
InChI Key BEQVQKJCLJBTKZ-UHFFFAOYSA-N
Synonym phosphinic acid, diphenyl, hydroxydiphenylphosphine oxide, diphenylphosphinicacid, diphenyl phosphinic acid, diphenyl-phosphinic acid, acmc-1boli, di phenyl phosphinic acid, p,p-diphenylphosphinic acid, ksc180o2h, phosphinicacid, p,p-diphenyl
PubChem CID 15567
ChEBI CHEBI:37832
IUPAC Name diphenylphosphinic acid
SMILES OP(=O)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 193.0°C to 196.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula (C6H5)2P(O)OH
Quantity 5 g
Beilstein 16, 791
Formula Weight 218.19
Percent Purity 99%
Physical Form Fine Crystalline Solid
Chemical Name or Material Diphenylphosphinic acid, 99%
Show More Show Less
EINECSNumber 216-948-5
RTECSNumber SZ5315000

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.