Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diphenylsilane, 97%

Catalog No. p-123038
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle

CAS: 775-12-2 | C12H12Si | 184.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 775-12-2
Molecular Formula C12H12Si
Molecular Weight (g/mol) 184.31
MDL Number MFCD00003002
InChI Key BPYFPNZHLXDIGA-UHFFFAOYSA-N
Synonym diphenylsilane, silane, diphenyl, diphenylsilicon, benzene, 1,1'-silylenebis, diphenyl silane, di phenyl silicon, diphenylsilylene radical, ph 2sih2
PubChem CID 6327659
IUPAC Name cyclohexa-2,5-dien-1-ylidene(phenyl)silanide
SMILES C1=CC=C(C=C1)[Si-]=C2C=C[CH+]C=C2

Specifications

Color Colorless
Density 0.9900g/mL
Boiling Point 95°C to 97°C (13.0 mmHg)
Flash Point 98°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Refractive Index 1.5780 to 1.58
Linear Formula (C6H5)2SiH2
Quantity 5 g
Fieser 01,348; 13,153; 15,151; 16,160; 17,132
Specific Gravity 0.99
Solubility Information Solubility in water: insoluble in water
Formula Weight 184.31
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Diphenylsilane, 97%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 212-271-4
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.