Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diphenylsilanediol, 98%

Catalog No. p-7080794
Click to view available options
Quantity:
100 g

CAS: 947-42-2 | C12H12O2Si | 216.311 g/mol

Diphenylsilanediol is used in Medicine field, electronics industry and polymer material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diphenylsilanediol is used in Medicine field, electronics industry and polymer material.

Solubility
Reacts with water.

Notes
Stable under normal temperatures and pressures. Incompatible with Oxidizing agents. Store at room temperature. Air & Light Sensitive.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 947-42-2
Molecular Formula C12H12O2Si
Molecular Weight (g/mol) 216.311
MDL Number MFCD00002101
InChI Key OLLFKUHHDPMQFR-UHFFFAOYSA-N
Synonym diphenylsilanediol, silanediol, diphenyl, diphenyldihydroxysilane, difenyl-dihydroxysilan, silane, dihydroxydiphenyl, dihydroxy diphenyl silane, diphenylsilandiol, silanediol, 1,1-diphenyl, unii-36x37p8nbb, difenyl-dihydroxysilan czech
PubChem CID 13693
IUPAC Name dihydroxy(diphenyl)silane
SMILES C1=CC=C(C=C1)[Si](C2=CC=CC=C2)(O)O

Specifications

Melting Point ∼150°C (decomposition)
Quantity 100 g
Linear Formula (C6H5Si(OH)2
UN Number UN1325
Beilstein 2523445
Solubility Information Reacts with water.
Formula Weight 216.31
Percent Purity 98%
Odor Odorless
Chemical Name or Material Diphenylsilanediol
Hazard Category H228-H315-H319-H335
Hazard Statement GHS H Statement
H228
Flammable solid.
Precautionary Statement P210-P240-P241-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
DOTInformation Hazard Class: 4.1; Packaging Group: II
EINECSNumber 213-427-4
RTECSNumber VV3640000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.