Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diphenylsulfone-3,3'-disulfonic Acid Disodium Salt 97.0+%, TCI America™
SDP

Catalog No. D091625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D091625G 25 g
D09165G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D091625G Supplier TCI America Supplier No. D091625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 39616-93-8
Molecular Formula C12H8Na2O8S3
Molecular Weight (g/mol) 422.348
MDL Number MFCD00059845
InChI Key GOPLREGDBGPRSC-UHFFFAOYSA-L
PubChem CID 18324062
IUPAC Name disodium;3-(3-sulfonatophenyl)sulfonylbenzenesulfonate
SMILES C1=CC(=CC(=C1)S(=O)(=O)[O-])S(=O)(=O)C2=CC(=CC=C2)S(=O)(=O)[O-].[Na+].[Na+]

Specifications

Color White-Yellow
Quantity 25 g
Synonym Disodium Diphenylsulfone-3,3′-disulfonate
Formula Weight 422.35
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Diphenylsulfone-3,3′-disulfonic Acid Disodium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.