Learn More
Specifications
Specifications
| Color | Black-Green |
| Boiling Point | 61°C |
| Melting Point | -64°C |
| CAS Max % | 98.9901, 0.9999, 0.01 |
| Linear Formula | C13H12N4S |
| UN Number | UN1888 |
| Density | 1,485kg/m3 |
| Solubility Information | Soluble in water |
| Formula Weight | 256.3 |
| Identification | Passes Test |
| Show More |
Safety and Handling
| Hazard Category | Warning |
| Hazard Statement | GHS H Statement Harmful if swallowed. Suspected of causing cancer. Toxic to aquatic life. |
| Precautionary Statement | GHS P Statement Obtain special instructions before use. Do not handle until all safety precautions have been read and understood. Do not eat, drink or smoke when using this product. Wear protective gloves, protective clothing, eye protection, face protection. Wash exposed skin thoroughly after handling. Avoid release to the environment. If exposed or concerned: Get medical advice/attention. If swallowed: Rinse mouth. Call a poison center or doctor if you feel unwell. Store locked up. Dispose of contents/container to comply with local, state and federal regulations. |
| Recommended Storage | Room Temperature |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.