Learn More
Diphenylzinc, 98+%, Thermo Scientific Chemicals
Description
Diphenylzinc is used as the synthetic equivalent of ph- synthon. It is also useful in a Michael 1,4-addition reactions. It is used as a catalyst for propylene oxide polymerization in benzene medium. Further, it is a reactive coupling compound in Negishi cross-coupling reactions. In addition to this, it utilized in the preparation of arylcopper, arylsilver and arylgold compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 107°C |
Boiling Point | 280°C to 285°C |
Quantity | 5 g |
UN Number | UN3181 |
Sensitivity | Air and moisture sensitive |
Solubility Information | Soluble in most organic solvents. |
Formula Weight | 219.58 |
Percent Purity | ≥98% |
Physical Form | Crystalline |
Chemical Name or Material | Diphenylzinc |
Safety and Handling
Hazard Category | H250-H314 |
Hazard Statement | GHS H Statement H228 Flammable solid. |
Precautionary Statement | P210-P222-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P335+P334-P363-P370+P378q-P501c |
DOTInformation | Hazard Class: 4.2; Packaging Group: I |
EINECSNumber | 214-082-2 |
TSCA | No |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.