Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Diphtheria Toxin Polyclonal Antibody, Biotin
GREENER_CHOICE

Catalog No. PIPA173139
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA173139 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA173139 Supplier Invitrogen™ Supplier No. PA173139
Add to Cart
Add to Cart

Goat Polyclonal Antibody

The PA1-73139 antibody reacts with Diphtheria toxin. PA1-73139 has been successfully used in Immunohistochemical, ELISA, Immunofluorescence and Western blot applications. The PA1-73139 antibody was raised against purified diphtheria toxoid.

Diphtheria toxin is a 58 kDa protein secreted by lysogenic strains of Corynebacterium diphtheriae. The toxin causes the disease diphtheria in humans by gaining entry into the cell cytoplasm and inhibiting protein synthesis. The mechanism of inhibition involves transfer of the ADP-ribose group of NAD to elongation factor-2 (EF-2), rendering EF-2 inactive. The catalysed reaction is as follows: NAD + + peptide diphthamide = nicotinamide + peptide N-(ADP-D-ribosyl)diphthamide The crystal structure of the diphtheria toxin homodimer has been determined to 2.5A resolution. The structure reveals a Y-shaped molecule of 3 domains, a catalytic domain (fragment A), whose fold is of the alpha + beta type; a transmembrane (TM) domain, which consists of 9 alpha-helices, 2 pairs of which may participate in pH-triggered membrane insertion and translocation; and a receptor-binding domain, which forms a flattened beta-barrel with a jelly-roll-like topology. The TM- and receptor binding-domains together constitute fragment B.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Diphtheria Toxin
Applications ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 5.0 mg/mL
Conjugate Biotin
Formulation PBS with 0.1% sodium azide; pH 7.2
Gene Alias DT
Host Species Goat
Immunogen Purified diphtheria toxoid.
Purification Method IgG fraction
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.