Learn More
Invitrogen™ Diphtheria Toxin Polyclonal Antibody, HRP

Description
The PA1-73138 antibody reacts with Diphtheria toxin. PA1-73138 has been successfully used in ELISA applications. The PA1-73138 antibody was raised against purified diphtheria toxoid.
Specifications
Specifications
| Antigen | Diphtheria Toxin |
| Applications | ELISA |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | HRP |
| Formulation | PBS with 10mg/mL BSA and 0.002% thimerosal |
| Gene Alias | DT |
| Host Species | Goat |
| Immunogen | Purified diphtheria toxoid. |
| Purification Method | IgG fraction |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.