Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Diphtheria Toxoid Antibody (17.6B9), Janelia Fluor™ 646, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP223596JF646
Description
Diphtheria Toxoid Monoclonal antibody specifically detects Diphtheria Toxoid in Bacteria samples. It is validated for ELISASpecifications
| Diphtheria Toxoid | |
| Monoclonal | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Bacteria | |
| Purified |
| ELISA | |
| 17.6B9 | |
| ELISA | |
| C. Diphtheria toxin, Corynebacterium diphtheriae toxin, Corynebacterium diphtheriae toxoid, DIP0222, Diptheria toxin, Diptheriae toxin, DTx | |
| Diphtheria toxoid prepared from toxin purified from C. diphtheriae subsp. intermedius Park-Williams no. 8 by formaldehyde inactivation (2), and adsorbed to aluminum hydroxide gel. | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction