Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(-)-Dipivaloyl-L-tartaric Acid 98.0+%, TCI America™
SDP

Catalog No. D268625G
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 65259-81-6
Molecular Formula C14H22O8
Molecular Weight (g/mol) 318.32
MDL Number MFCD00066535
InChI Key UFHJEZDFEHUYCR-YZYOREDDNA-N
Synonym +-dipivaloyl-d-tartaric acid, 2s,3s-2,3-bis pivaloyloxy succinic acid, d-+-dipivaloyl tartaric acid, pubchem8173, --bis 2,2-dimethylpropionyl-l-tartaric acid, dipivaloyl-l-tartaric acid, +-dipivaloyl-d-tartaricacid, +-dpta, ?-o,o'-di-pivaloyl-l-tartaric acid, +-o,o'-di-pivaloyl-d-tartaric acid
PubChem CID 16212279
IUPAC Name (2R,3R)-2,3-bis[(2,2-dimethylpropanoyl)oxy]butanedioic acid
SMILES CC(C)(C)C(=O)O[C@H]([C@@H](OC(=O)C(C)(C)C)C(O)=O)C(O)=O

Specifications

Melting Point 150°C
Color White
Quantity 25 g
Formula Weight 318.32
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material (-)-Dipivaloyl-L-tartaric Acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.