Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dipropylamine, 99%

Catalog No. p-123433
Click to view available options
Quantity:
250 mL
1 L
Packaging:
Glass bottle

Catalog No. AC117492500

Supplier: Thermo Scientific Chemicals 117492500

Only null left
Add to Cart

CAS: 142-84-7 | C6H15N | 101.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 142-84-7
Molecular Formula C6H15N
Molecular Weight (g/mol) 101.19
MDL Number MFCD00009362
InChI Key WEHWNAOGRSTTBQ-UHFFFAOYSA-N
Synonym dipropylamine, di-n-propylamine, 1-propanamine, n-propyl, n-dipropylamine, n-propyl-1-propanamine, rcra waste number u110, dipropyl amine, rcra waste no. u110, unii-60p318iiry, ccris 4805
PubChem CID 8902
IUPAC Name N-propylpropan-1-amine
SMILES CCCNCCC

Specifications

Melting Point -63.0°C
Density 0.7400g/mL
Boiling Point 105.0°C to 110.0°C
Flash Point 7°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4030 to 1.4050
Linear Formula (CH3CH2CH2)2NH
Quantity 250 mL
Beilstein 04, 138
Specific Gravity 0.74
Merck Index 15, 3383
Solubility Information Solubility in water: 46g/L (20°C). Other solubilities: freely soluble in alcohol
Viscosity 0.5 mPa.s (20°C)
Formula Weight 101.19
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Dipropylamine, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
Toxic if inhaled.
Toxic in contact with skin.
May cause respiratory irritation.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rin
EINECSNumber 205-565-9
RTECSNumber JL9200000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.