Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dipropylene Glycol Diacrylate (stabilized with MEHQ) 80.0+%, TCI America™
SDP

Catalog No. D3470100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D3470100G 100 g
D3470500G 500 g
D347025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D3470100G Supplier TCI America Supplier No. D3470100G

Chemical Identifiers

CAS 57472-68-1
Molecular Formula C12H18O5

Specifications

Color Yellow
Quantity 100 g
Formula Weight 242.27
Percent Purity ≥80.0% (GC)
Physical Form Liquid
Chemical Name or Material Dipropylene Glycol Diacrylate (stabilized with MEHQ)
EINECSNumber (7)-0152
RTECSNumber UD3643950
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.