Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Dipyridamole
GREENER_CHOICESDP

Catalog No. 0691500
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06-915-00 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 06-915-00 Supplier Tocris Bioscience™ Supplier No. 0691/500

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Adenosine transport inhibitor

Coronary vasodilator; adenosine transport inhibitor. Phosphodiesterase inhibitor (IC50 values are 0.37, 0.38, 0.45, 0.9 and 4.5 μM for PDE11, 6, 10, 5 and 8 respectively). Inhibits ENT1 and ENT2 (IC50 = 144.8 nM and Ki = 8.18 nM for ENT1).

Chemical Identifiers

CAS 58-32-2
Molecular Formula C24H40N8O4

Specifications

Quantity 500 mg
Formula Weight 504.63
Purity >99%
Molecular Weight (g/mol) 504.636
Chemical Name or Material Dipyridamole
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.