Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DIRAS1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5104509
Description
Antibody detects endogenous levels of total DIRA1.
DIRAS1 belongs to a distinct branch of the functionally diverse Ras superfamily of monomeric GTPases.
Specifications
DIRAS1 | |
Polyclonal | |
Unconjugated | |
Diras1 | |
DIRAS family GTP binding RAS like 1; DIRAS family GTPase 1; DIRAS family, GTP-binding RAS-like 1; DIRAS1; Di-Ras1; Distinct subgroup of the Ras family member 1; GBTS1; GTP-binding protein Di-Ras1; ras-related inhibitor of cell growth; RIG; small GTP-binding tumor suppressor 1 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
148252, 208666 | |
-20°C | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
O95057, Q91Z61 | |
Diras1 | |
A synthesized peptide derived from human DIRAS1(Accession O95057), corresponding to amino acid residues F161-M198. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction