Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DIRAS2 Antibody (OTI10H1), Alexa Fluor™ 532, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271967AF532
Description
DIRAS2 Monoclonal specifically detects DIRAS2 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
DIRAS2 | |
Monoclonal | |
Alexa Fluor 532 | |
50 mM sodium borate with 0.05% sodium azide | |
DIRAS2 | |
Recombinant protein fragment corresponding to amino acids 1-196 of human DIRAS2 (NP_060064) produced in E.coli. | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Western Blot | |
OTI10H1 | |
Western Blot | |
DIRAS family, GTP-binding RAS-like 2, Di-Ras2, Distinct subgroup of the Ras family member 2, DKFZp761C07121, GTP-binding protein Di-Ras2 | |
Mouse | |
0.1 mL | |
54769 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction