Learn More
Invitrogen™ DIRAS2 Monoclonal Antibody (OTI10G5)

Description
DIRAS2 Monoclonal Antibody for Western Blot, IHC (P)
Specifications
Specifications
| Antigen | DIRAS2 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | OTI10G5 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | DIRAS2 |
| Gene Accession No. | Q96HU8 |
| Gene Alias | 2900052J15Rik; AI414999; DIRAS family GTP binding RAS like 2; DIRAS family GTPase 2; DIRAS family, GTP-binding RAS-like 2; DIRAS2; Di-Ras2; Distinct subgroup of the Ras family member 2; GTP-binding protein Di-Ras2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.