Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DIRAS2 Mouse anti-Human, Clone: OTI10H1, lyophilized, TrueMAB™
SDP

Catalog No. 501669487
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-166-9487 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-166-9487 Supplier OriGene Supplier No. CF809430
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO

DIRAS2 belongs to a distinct branch of the functionally diverse Ras superfamily of monomeric GTPases.

Specifications

Antigen DIRAS2
Applications Western Blot
Classification Monoclonal
Clone OTI10H1
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene DIRAS2
Gene Accession No. Q96HU8
Gene Alias Di-Ras2
Gene Symbols DIRAS2
Host Species Mouse
Immunogen Human recombit protein fragment corresponding to amino acids 1-196 of human DIRAS2 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54769
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.