Learn More
DIRAS2 Mouse anti-Human, Clone: OTI9G7, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF809400

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
DIRAS2 belongs to a distinct branch of the functionally diverse Ras superfamily of monomeric GTPases.Specifications
DIRAS2 | |
Monoclonal | |
Unconjugated | |
DIRAS2 | |
Di-Ras2 | |
Mouse | |
Affinity Chromatography | |
RUO | |
54769 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI9G7 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q96HU8 | |
DIRAS2 | |
Human recombit protein fragment corresponding to amino acids 1-196 of human DIRAS2 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.