Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DIRAS2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA571083
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 93%.
DIRAS2 belongs to a distinct branch of the functionally diverse Ras superfamily of monomeric GTPases.
Specifications
DIRAS2 | |
Polyclonal | |
Unconjugated | |
DIRAS2 | |
2900052J15Rik; AI414999; DIRAS family GTP binding RAS like 2; DIRAS family GTPase 2; DIRAS family, GTP-binding RAS-like 2; DIRAS2; Di-Ras2; Distinct subgroup of the Ras family member 2; GTP-binding protein Di-Ras2 | |
Rabbit | |
Affinity chromatography | |
RUO | |
54769 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q96HU8 | |
DIRAS2 | |
synthetic peptide directed towards the C-terminal region of human DIRAS2. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction