Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DIRAS2 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982534
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2534 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2534 Supplier Bioss Supplier No. BS13003R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DIRAS2 belongs to a distinct branch of the functionally diverse Ras superfamily of monomeric GTPases.

Specifications

Antigen DIRAS2
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene DIRAS2
Gene Accession No. Q96HU8
Gene Alias 2900052J15Rik; AI414999; BOS_9208; DIRAS family GTP binding RAS like 2; DIRAS family GTPase 2; DIRAS family, GTP-binding RAS-like 2; Diras2; Di-Ras2; Di-Ras2-like; Distinct subgroup of the Ras family member 2; GTP-binding protein Di-Ras2; LOC100352632
Gene Symbols DIRAS2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human DIRAS2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54769
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.