Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DIRAS3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA555872
Description
Immunogen sequence: SDDTHREVAL NDGATCAMEW NCAFMEISAK TDVNVQELFH MLLNYKKKPT TGLQEPEKKS QMPNTTEKLL DKCIIM Highest antigen sequence identity to the following orthologs: Mouse - 45%, Rat - 45%.
This gene is a member of the ras superfamily, and is expressed in normal ovarian and breast epithelial cells, but not in ovarian and breast cancers. It is an imprinted gene, with monoallelic expression of the paternal allele, which is associated with growth suppression. Thus, this gene appears to be a putative tumor suppressor gene whose function is abrogated in ovarian and breast cancers.
Specifications
DIRAS3 | |
Polyclonal | |
Unconjugated | |
DIRAS3 | |
ARHI; DIRAS family GTP binding RAS like 3; DIRAS family GTPase 3; DIRAS family, GTP-binding RAS-like 3; DIRAS3; Distinct subgroup of the Ras family member 3; GTP-binding protein Di-Ras3; NOEY2; ras homolog gene family, member I; RGD1565168; RHOI; rho-related GTP-binding protein RhoI | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
9077 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O95661 | |
DIRAS3 | |
Recombinant protein corresponding to Human DIRAS3. Recombinant protein control fragment (Product #RP-94376). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction