Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DIRAS3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596877
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596877 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596877 Supplier Invitrogen™ Supplier No. PA596877
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MGNASFGSKE QKLLKRLRLL PALLILRAFK PHRKIRDYRV VVVGTAGVGK STLLHKWASG NFRHEYLPTI ENTYCQLLGC SHGVLSLHIT DSKSGDGNRA LQRHVIARGH AFVLVYSVTK KETLEELKAF YELICKIKGN NLHKFPIVLV GNKSDDTHRE VALNDGATCA MEWNCAFMEI SAKTDVNVQE LFHMLLNYKK KPTTGLQEPE KKSQMPNTTE KLLDKCIIM; Positive Samples: NIH/3T3, Mouse liver, Mouse kidney, Rat liver; Cellular Location: Cell membrane, Cytoplasmic side, Lipid-anchor.

This gene is a member of the ras superfamily, and is expressed in normal ovarian and breast epithelial cells, but not in ovarian and breast cancers. It is an imprinted gene, with monoallelic expression of the paternal allele, which is associated with growth suppression. Thus, this gene appears to be a putative tumor suppressor gene whose function is abrogated in ovarian and breast cancers.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DIRAS3
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.71 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene DIRAS3
Gene Accession No. O95661
Gene Alias ARHI; DIRAS family GTP binding RAS like 3; DIRAS family GTPase 3; DIRAS family, GTP-binding RAS-like 3; DIRAS3; Distinct subgroup of the Ras family member 3; GTP-binding protein Di-Ras3; NOEY2; ras homolog gene family, member I; RGD1565168; RHOI; rho-related GTP-binding protein RhoI
Gene Symbols DIRAS3
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-229 of human DIRAS3 (NP_0046661).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 366733
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.