Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DIRC2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100787
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100787 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100787 Supplier Invitrogen™ Supplier No. PA5100787
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total DIRC2.

Electrogenic metabolite transporter.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DIRC2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Dirc2
Gene Accession No. Q8BFQ6, Q96SL1
Gene Alias Dirc2; Disrupted in renal cancer protein 2; disrupted in renal carcinoma 2; disrupted in renal carcinoma 2 (human); disrupted in renal carcinoma 2 homolog; disrupted in renal carcinoma protein 2; Disrupted in renal carcinoma protein 2 homolog; RCC4; renal cell carcinoma 4; SLC49A4; Solute carrier family 49 member 4
Gene Symbols Dirc2
Host Species Rabbit
Immunogen A synthesized peptide derived from human DIRC2(Accession Q96SL1), corresponding to amino acid residues F198-A248.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 224132, 84925
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.